Ammonium cerium (IV) sulfate dihydrate

Synonyms: Tetraammonium cerium tetrakis(sulphate) dihydrate, Cerium (IV) ammonium sulphate dihydrate, Tetraammonium cerium(IV) sulphate dihydrate, Ammonium cerium(IV) sulphate hydrate, Ammonium ceric sulphate dihydrate

221759-100G 221759-500G
220044-529EA 180 USD
220044-529 220044-530
Ammonium cerium (IV) sulfate dihydrate
Ammonium cerium(IV) sulphate dihydrate
Formula: (NH₄)₂Ce(SO₄)₄·2H₂O
MW: 632.55 g/mol
Melting Pt: 130 °C
MDL Number: MFCD00149417
CAS Number: 10378-47-9
EINECS: 231-567-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR