Ammonium iron (III) sulfate dodecahydrate ACS

Warning

221260-2.5KG 221260-25G 221260-500G
220044-805EA 326.15 USD
220044-805 220044-806 220044-807
Ammonium iron (III) sulfate dodecahydrate ACS
Ammonium iron(III) sulphate dodecahydrate
Formula: NH₄Fe(SO₄)₂·12H₂O
Boiling Pt: 230 °C (1013 hPa)
Melting Pt: 39…41 °C
Density: 1.71 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150004
CAS Number: 7783-83-7
EINECS: 233-382-4
Merck Index: 12,00549

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR