(S)-(-)-1-Phenylethyl isocyanate ≥99%

Synonyms: (S)-(-)-α-Phenethyl isocyanate, (S)-(-)-α-Methylbenzyl isocyanate, (S)-1-phenylethylisocyanate, (S)-(-)-1-Phenethyl isocyanate

220566-1G 220566-5G
220043-823EA 76.46 USD
220043-823 220043-824
(S)-(-)-1-Phenylethyl isocyanate ≥99%
(S)-(-)-1-Phenylethyl isocyanate
Formula: C₉H₉NO
MW: 147.18 g/mol
Boiling Pt: 55 °C (1013 hPa)
Density: 1.05 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00002036
CAS Number: 14649-03-7
EINECS: 238-698-6

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR