1-Amino-2-methylbutane ≥97%

Synonyms: 2-Methylbutylamine, 2-Methyl-1-butylamine

220523-25G 220523-5G
220043-818EA 641.85 USD
220043-818 220043-819
1-Amino-2-methylbutane ≥97%
1-Amino-2-methylbutane
Formula: C₅H₁₃N
MW: 87.16 g/mol
Boiling Pt: 97 °C (1013 hPa)
Density: 0.755 g/cm³ (18 °C)
MDL Number: MFCD00008147
CAS Number: 96-15-1
EINECS: 202-483-5

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR