Aluminium isopropoxide ≥98%

Danger

Synonyms: Aluminiumtriisopropylate, Aluminiumisopropylate, Aluminiumtriisopropoxide, Aluminium isopropoxide

220418-100G 220418-1KG
220044-725EA 72 USD
220044-725 220044-726
Aluminium isopropoxide ≥98%
Aluminium isopropoxide
Formula: C₉H₂₁AlO₃
MW: 204.25 g/mol
Boiling Pt: 135 °C (13 hPa)
Melting Pt: 119 °C
Density: 0.9771 g/cm³ (20 °C)
MDL Number: MFCD00008870
CAS Number: 555-31-7
EINECS: 209-090-8
UN: 3181
ADR: 4.1,II
Merck Index: 13,00346

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR