3-Hydroxy-p-anisic acid ≥97%

Warning

Synonyms: 3-Hydroxy-4-methoxybenzoic acid, Isovanillic acid

220108-25G 220108-5G
220044-517EA 20.31 USD
220044-517 220044-518
3-Hydroxy-p-anisic acid ≥97%
3-Hydroxy-p-anisic acid
Formula: C₈H₈O₄
MW: 168.15 g/mol
Boiling Pt: 327 °C (1013 hPa)
Melting Pt: 258…259 °C
MDL Number: MFCD00002507
CAS Number: 645-08-9
EINECS: 211-430-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR