3-(Indol-3-yl)propionic acid

Synonyms: Indole-3-propionic acid, 3-Indolepropionic acid

220027-1G 220027-10G
220044-510EA 31.77 USD
220044-510 220044-509
3-(Indol-3-yl)propionic acid
3-(Indol-3-yl)propionic acid
Formula: C₁₁H₁₁NO₂
MW: 189.21 g/mol
Melting Pt: 129…132 °C
Density: 1.356 g/cm³ (20 °C)
MDL Number: MFCD00005660
CAS Number: 830-96-6
EINECS: 212-600-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR