1-Fluoro-4-iodobenzene ≥99%

Warning

Synonyms: 4-Fluoroiodobenzene

219428-5G 219428-50G
220044-653EA 39.85 USD
220044-653 220044-652
1-Fluoro-4-iodobenzene ≥99%
1-Fluoro-4-iodobenzene
Formula: C₆H₄FI
MW: 222 g/mol
Boiling Pt: 182 °C (1013 hPa)
Melting Pt: -18 °C
Density: 1.9253 g/cm³ (15 °C)
MDL Number: MFCD00001052
CAS Number: 352-34-1
EINECS: 206-522-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR