N,N-Dimethyl-1,3-phenylenediammonium dichloride ≥99%

Synonyms: N1,N1-Dimethylbenzene-1,3-diamine dihydrochloride, N,N-Dimethyl-1,3-phenylenediamine dihydrochloride, N,N-Dimethyl-m-phenylenediamine dihydrochloride

219223-25G 219223-5G
220043-944EA 211.08 USD
220043-944 220044-635
N,N-Dimethyl-1,3-phenylenediammonium dichloride ≥99%
N,N-Dimethyl-1,3-phenylenediammonium dichloride
Formula: C₈H₁₄Cl₂N₂
MW: 209.12 g/mol
Melting Pt: 218 °C
MDL Number: MFCD00012971
CAS Number: 3575-32-4
EINECS: 222-693-0
UN: 2811
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR