5-Bromopyrimidine

Warning

219142-25G 219142-5G
220043-936EA 88.31 USD
220043-936 220043-937
5-Bromopyrimidine
5-Bromopyrimidine
Formula: C₄H₃BrN₂
MW: 158.99 g/mol
Boiling Pt: 168 °C (727 mmHg)
Melting Pt: 75…76 °C
MDL Number: MFCD00006117
CAS Number: 4595-59-9
EINECS: 224-992-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR