β-Butyrolactone

Synonyms: beta-Methylpropiolactone, beta-Butyrolactone, 1,3-Butyrolactone, 4-Methyl-2-oxetanone, 4-Methyloxetan-2-one, Butan-3-olide

219126-50G 219126-10G
220043-935EA 78.18 USD
220043-935 220043-934
β-Butyrolactone
β-Butyrolactone
Formula: C₄H₆O₂
MW: 86.09 g/mol
Boiling Pt: 163 °C (1013 hPa)
Melting Pt: –43.5 °C
Density: 1.0555 g/cm³ (20 °C)
MDL Number: MFCD00005170
CAS Number: 3068-88-0
EINECS: 221-330-3
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR