D-(-)-Aspartic acid ≥99%

Synonyms: D-Aminosuccinic acid, D(-)-2-Amino-1,4-butanedioic acid, (R)-(-)-2-Amino-1,4-butanedioic acid, (R)-(-)-Aspartic acid, (R)-()-Aminosuccinic acid, (R)-Aminosuccinic acid, (R)-2-Aminosuccinic acid, D-Asparaginic acid, D-Aspartic acid, H-D-Asp-OH

219096-25G 219096-100G
220043-933EA 174.46 USD
220043-933 220043-932
D-(-)-Aspartic acid ≥99%
D(-)-Aspartic acid
Formula: C₄H₇NO₄
MW: 133.1 g/mol
Melting Pt: 251 °C
MDL Number: MFCD00063081
CAS Number: 1783-96-6
EINECS: 217-234-6
Merck Index: 13,00845

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR