1,1'-Carbonyldiimidazole

Danger

Synonyms: Di(1H-imidazol-1-yl)methanone, CDI Crosslinker, 1,1'-Carbonylbis(imidazole), 1,1'-Carbonylbis-1H-imidazole, N,N'-Carbonyldiimidazole, CDI

21860-5G 21860-25G 21860-100G
220043-197EA 35.65 USD
220043-197 220043-196 220043-195
1,1'-Carbonyldiimidazole
1,1'-Carbonyldiimidazole
Formula: C₇H₆N₄O
MW: 162.15 g/mol
Melting Pt: 111…116 °C
Storage Temperature: Ambient
MDL Number: MFCD00005286
CAS Number: 530-62-1
EINECS: 208-488-9
UN: 3263
ADR: 8,II
Merck Index: 14,01819

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR