(±)-3-Octanol ≥99%

Warning

Synonyms: 3-Octanol, (±)-Ethyl pentyl carbinol, (±)-3-Hydroxy octane, (±)-Pentyl ethyl carbinol, DL-3-Octanol

218405-250G 218405-50G
220043-190EA 178.77 USD
220043-190 220043-191
(±)-3-Octanol ≥99%
(±)-3-Octanol
Formula: C₈H₁₈O
MW: 130.23 g/mol
Boiling Pt: 174 °C (1013 hPa)
Melting Pt: –45 °C
Density: 0.819 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004590
CAS Number: 589-98-0
EINECS: 209-667-4

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR