Cesium hydrogen carbonate ≥99.9%

218227-100G 218227-25G
220044-625EA 222.92 USD
220044-625 220044-626
Cesium hydrogen carbonate ≥99.9%
Cesium hydrogen carbonate
Formula: CHCsO₃
MDL Number: MFCD00010961
CAS Number: 15519-28-5
EINECS: 239-554-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR