Lithium iodide ≥99%

218219-10G 218219-250G 218219-50G
220044-621EA 203.08 USD
220044-621 220044-622 220044-623
Lithium iodide ≥99%
Lithium iodide
Formula: ILi
MW: 133.85 g/mol
Boiling Pt: 1177 °C (1013 hPa)
Melting Pt: 450 °C
Density: 3.49 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011092
CAS Number: 10377-51-2
EINECS: 233-822-5
Merck Index: 13,05557

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR