Tetrabutylammonium tetrafluoroborate

Warning

Synonyms: Tetrabutylammoniumtetrafluoroborate

217964-25G 217964-100G 217964-5G
220044-605EA 126 USD
220044-605 220044-604 220044-606
Tetrabutylammonium tetrafluoroborate
Tetrabutylammonium tetrafluoroborate
Formula: C₁₆H₃₆BF₄N
MW: 329.27 g/mol
Melting Pt: 159…162 °C
Storage Temperature: Ambient
MDL Number: MFCD00011634
CAS Number: 429-42-5
EINECS: 207-058-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR