Tin(IV) chloride ≥99.995%

Danger

Synonyms: Stannic chloride, Tin tetrachloride

217913-100G 217913-25G 217913-5G
220044-598EA 218.08 USD
220044-598 220044-599 220044-600
Tin(IV) chloride ≥99.995%
Tin(IV) chloride
Formula: Cl₄Sn
MW: 260.52 g/mol
Boiling Pt: 114 °C (1013 hPa)
Melting Pt: –33 °C
Density: 2.22 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011242
CAS Number: 7646-78-8
EINECS: 231-588-9
UN: 1827
ADR: 8,II
Merck Index: 12,08929

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR