Iron(III) bromide

Warning

Synonyms: Ferric bromide, Iron tribromide

217883-10G 217883-50G
220044-593EA 155.38 USD
220044-593 220044-594
Iron(III) bromide
Iron(III) bromide
Formula: Br₃Fe
MW: 295.56 g/mol
MDL Number: MFCD00016082
CAS Number: 10031-26-2
EINECS: 233-089-1
UN: 3260
ADR: 8,III
Merck Index: 12,04060

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR