2-Bromo-5-(trifluoromethyl)aniline ≥97%

Danger

Synonyms: 3-Amino-4-bromobenzotrifluoride, 6-Bromo-α,α,α-trifluoro-m-toluidine, 3-Amino-4-bromo-α,α,α-trifluorotoluene, 3-Amino-4-brom-α,α,α-trifluortoluol

217867-5G 217867-1G
220044-592EA 66.55 USD
220044-592 220044-591
2-Bromo-5-(trifluoromethyl)aniline ≥97%
2-Bromo-5-(trifluoromethyl)aniline
Formula: C₇H₅BrF₃N
MW: 240.02 g/mol
Boiling Pt: 94 - 96 °C (12 -rr)
Melting Pt: 47 - 49 °C
Density: 1.694 g/cm³ (25 °C)
MDL Number: MFCD00007637
CAS Number: 454-79-5
EINECS: 207-227-6
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR