5-Amino-4,6-dichloropyrimidine

217735-25G 217735-5G
220044-583EA 313.38 USD
220044-583 220044-584
5-Amino-4,6-dichloropyrimidine
5-Amino-4,6-dichloropyrimidine
Formula: C₄H₃Cl₂N₃
MW: 163.99 g/mol
Boiling Pt: 262 °C (1013 hPa)
Melting Pt: 147 °C
MDL Number: MFCD00006108
CAS Number: 5413-85-4
EINECS: 226-503-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR