Trichloroacetyl isocyanate ≥96%

217328-1G 217328-10G
220044-482EA 58.58 USD
220044-482 220044-481
Trichloroacetyl isocyanate ≥96%
Trichloroacetyl isocyanate
Formula: C₃Cl₃NO₂
MW: 188.4 g/mol
Boiling Pt: 80…85 °C (27 hPa)
Density: 1.59 g/cm³ (20 °C)
Flash Pt: 65 °C
MDL Number: MFCD00002033
CAS Number: 3019-71-4
EINECS: 221-165-7
UN: 2927
ADR: 6.1,II

Order Now

Specification Test Results

Assay 96%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR