Sodium thiosulfate pentahydrate ≥99.5% ACS

217247-25G 217247-2.5KG 217247-500G
220044-479EA 34.09 USD
220044-479 220044-478 220043-738
Sodium thiosulfate pentahydrate ≥99.5% ACS
Sodium thiosulphate pentahydrate
Formula: Na₂S₂O₃.5H₂O
MW: 248.19 g/mol
Melting Pt: 48 °C
Density: 1.67 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149186
CAS Number: 10102-17-7
EINECS: 231-867-5
Merck Index: 12,08844

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR