Yttrium(III) nitrate tetrahydrate

Warning

217239-10G 217239-50G
220044-476EA 390.77 USD
220044-476 220044-477
Yttrium(III) nitrate tetrahydrate
Yttrium(III) nitrate tetrahydrate
Formula: N₃O₉Y·4H₂O
MDL Number: MFCD00149938
CAS Number: 13773-69-8
EINECS: 233-802-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR