Ytterbium trinitrate pentahydrate ≥99.999%

Synonyms: Ytterbium (III) nitrate pentahydrate

217220-25G 217220-5G
220044-474EA 587.69 USD
220044-474 220044-475
Ytterbium trinitrate pentahydrate ≥99.999%
Ytterbium trinitrate pentahydrate
Formula: N₃O₉Yb·5H₂O
MW: 449.13 g/mol
MDL Number: MFCD00149883
CAS Number: 35725-34-9
EINECS: 237-384-6
UN: 1477
ADR: 5.1,III

Order Now

Specification Test Results

Assay 99.999%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR