Terbium trinitrate hexahydrate ≥99.999%

217212-2G 217212-10G
220044-473EA 212.31 USD
220044-473 220044-472
Terbium trinitrate hexahydrate ≥99.999%
Terbium trinitrate hexahydrate
Formula: N₃O₉Tb·6H₂O
Melting Pt: 89.3 °C
MDL Number: MFCD00149868
CAS Number: 13451-19-9
EINECS: 233-138-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR