6-Aminonicotinic acid

Synonyms: 6-Aminopyridine-3-carboxylic acid, 6-Amino-3-pyridinecarboxylic acid

216879-5G 216879-1G
220044-451EA 47.06 USD
220044-451 220044-450
6-Aminonicotinic acid
6-Aminonicotinic acid
Formula: C₆H₆N₂O₂
MW: 138.13 g/mol
Melting Pt: ? 300 °C
MDL Number: MFCD00006326
CAS Number: 3167-49-5
EINECS: 221-630-4
Merck Index: 13,00455

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR