Magnesium bromide hexahydrate ≥99%

Warning

216844-250G 216844-1KG
220044-448EA 126.92 USD
220044-448 220044-447
Magnesium bromide hexahydrate ≥99%
Magnesium bromide hexahydrate
Formula: Br₂Mg·6H₂O
MW: 292.2 g/mol
Melting Pt: 146 °C
Density: 2 g/cm³ (20 °C)
MDL Number: MFCD00149780
CAS Number: 13446-53-2
EINECS: 232-170-9

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR