2,4-Toluenediisocyanate ≥80%, Technical Grade

Danger

Synonyms: 4-Methyl-m-phenylene diisocyanate, Tolylene 2,4-diisocyanate, 2,4-TDI

216836-1KG 216836-250G
220044-445EA 124.92 USD
220044-445 220044-446
2,4-Toluenediisocyanate ≥80%, Technical Grade
2,4-Toluenediisocyanate
Formula: C₉H₆N₂O₂
MW: 174.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00002011
CAS Number: 584-84-9
EINECS: 247-722-4
UN: 2078
ADR: 6.1,II

Order Now

Specification Test Results

Appearance (Color) Colorless or White
Appearance (Form) Conforms to Requirements (Liquid, Crystals or Solid)
Infrared spectrum Conforms to Structure
Tolylene-2,4-diisocyanate (GC) > 78.0 %
Product of Supplier Confirmed (BASF AG (Aktiengesellschaft)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR