Potassium chromate ≥99.0% ACS

Danger

216615-500G 216615-100G
220044-421EA 125.54 USD
220044-421 220044-420
Potassium chromate ≥99.0% ACS
Potassium chromate
Formula: K₂CrO₄
Boiling Pt: 1000 °C (1013 hPa)
Melting Pt: 985 °C
Density: 2.73 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011368
CAS Number: 7789-00-6
EINECS: 232-140-5
UN: 3087
ADR: 5.1,III
Merck Index: 12,07784

Order Now

Specification Test Results

Assay 99.0%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR