Boron trifluoride diethyl ether complex

Danger

Synonyms: Boron fluoride, ethyl ether complex, Boron trifluoride ethyl etherate, Boron trifluoride diethyl ether, Boron trifluoride etherate, Diethyl ether boron trifluoride, Boron trifluoride diethyl etherate

216607-100ML 216607-800ML
220044-418EA 54.06 USD
220044-418 220044-419
Boron trifluoride diethyl ether complex
Boron trifluoride diethyl ether complex
Formula: C₄H₁₀BF₃O
MW: 141.93 g/mol
Boiling Pt: 126 °C (1013 hPa)
Melting Pt: –60 °C
Density: 1.125 g/cm³ (25 °C)
Flash Pt: 59 °C
Storage Temperature: Ambient
MDL Number: MFCD00013194
CAS Number: 109-63-7
EINECS: 203-689-8
UN: 2604
ADR: 8,I
Merck Index: 13,01340

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR