Hydrazinium monochloride

Danger

Synonyms: Hydrazinium monochloride, Hydrazinium chloride, Hydrazine monohydrochloride, Hydrazine hydrochloride

216194-5G 216194-250G
220044-314EA 49.32 USD
220044-314 220044-313
Hydrazinium monochloride
Hydrazinium monochloride
Formula: H₅ClN₂
MW: 68.51 g/mol
Melting Pt: 89 - 93 °C
Density: 1.5 g/cm³ (20 °C)
MDL Number: MFCD00044368
CAS Number: 2644-70-4
EINECS: 220-154-4
UN: 2923
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR