Methyl-4-chlorobenzoate ≥99%

Synonyms: 4-Chlorobenzoic acid emthyl ester, Methyl 4-chlorobenzoate

216089-5G 216089-100G
220044-297EA 37.38 USD
220044-297 220044-296
Methyl-4-chlorobenzoate ≥99%
Methyl-4-chlorobenzoate
Formula: C₈H₇ClO₂
MW: 170.6 g/mol
Boiling Pt: 103…106 °C (15 mmHg)
Melting Pt: 43 °C
Density: 1.382 g/cm³ (20 °C)
MDL Number: MFCD00000621
CAS Number: 1126-46-1
EINECS: 214-420-9
Merck Index: 13,02144

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR