Lead(II) acetate trihydrate ≥99% ACS

Danger

Synonyms: Acetic acid lead (II) salt trihydrate, Lead diacetate trihydrate

215902-2.5KG 215902-500G 215902-25G
220043-729EA 235.69 USD
220043-729 220043-731 220043-730
Lead(II) acetate trihydrate ≥99% ACS
Lead(II) acetate trihydrate
Formula: (H₃CCOO)₂Pb·3H₂O
MW: 379.34 g/mol
Boiling Pt: 280 °C (1013 hPa)
Melting Pt: 75 °C
Density: 2.55 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150023
CAS Number: 6080-56-4
EINECS: 206-104-4
UN: 1616
ADR: 6.1,III
Merck Index: 13,05415
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=351511a44fbb598a2d7fa1fa4ab395427b6168f1&vid=67e858e5f0dc4d04dd796541b3c0ad5aa73f8dc8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR