Manganese(III) acetate dihydrate ≥97%

Synonyms: Acetic acid manganese (III) salt dihydrate, Manganese triacetate dihydrate

215880-25G 215880-100G 215880-5G
220043-725EA 292.31 USD
220043-725 220043-724 220043-726
Manganese(III) acetate dihydrate ≥97%
Manganese(III) acetate dihydrate
Formula: (H₃CCOO)₂Mn·2H₂O
MW: 209.06 g/mol
MDL Number: MFCD00150022
CAS Number: 19513-05-4
EINECS: 213-602-5

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR