Vanadium(IV) oxide

Synonyms: Vanadium dioxide

215821-5G 215821-20G
220043-723EA 206.15 USD
220043-723 220043-722
Vanadium(IV) oxide
Vanadium(IV) oxide
Formula: O₂V
MW: 82.94 g/mol
Boiling Pt: 2700 °C (1013 hPa)
Melting Pt: 238 °C
Density: 4.33 g/cm³ (20 °C)
MDL Number: MFCD00049702
CAS Number: 12036-21-4
EINECS: 234-841-1
UN: 3285
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR