1-Butyl-3-methylimidazolium methyl sulfate

Synonyms: [BMIM] [MeSO4], 1-n-Butyl-3-methylimidazolium methyl sulphate, ECOENG 411, BMIMMeSO4

83086-5G 83086-50G
220097-601EA 118.46 USD
220097-601 220097-600
1-Butyl-3-methylimidazolium methyl sulfate
1-Butyl-3-methylimidazolium methyl sulphate
Formula: C₉H₁₈N₂O₄S
MW: 250.32 g/mol
Density: 1.21 g/cm³ (20 °C)
MDL Number: MFCD03095437
CAS Number: 401788-98-5
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR