Copper(II) perchlorate hexahydrate

215392-5G 215392-100G
220044-232EA 48.31 USD
220044-232 220044-231
Copper(II) perchlorate hexahydrate
Copper(II) perchlorate hexahydrate
Formula: Cl₂CuO₈·6H₂O
Melting Pt: 82 °C
Density: 1.0491 g/cm³ (20 °C)
MDL Number: MFCD00149666
CAS Number: 10294-46-9
EINECS: 237-391-4
UN: 1481
ADR: 5.1,II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR