Rubidium chloride ≥99.8%

Synonyms: Rubidiumchloride

215260-50G 215260-10G
220044-215EA 458.46 USD
220044-215 220044-214
Rubidium chloride ≥99.8%
Rubidium chloride
Formula: RbCl
MW: 120.92 g/mol
Boiling Pt: 1390 °C (1013 hPa)
Melting Pt: 715 °C
Density: 2.8 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011187
CAS Number: 7791-11-9
EINECS: 232-240-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR