Tetrachlorosilane ≥99%

Warning

215120-1KG 215120-100G
220043-896EA 186.15 USD
220043-896 220043-895
Tetrachlorosilane ≥99%
Tetrachlorosilane
Formula: Cl₄Si
Boiling Pt: 57.6 °C (1013 hPa)
Melting Pt: –70 °C
Density: 1.48 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011229
CAS Number: 10026-04-7
EINECS: 233-054-0
UN: 1818
ADR: 8,II
Merck Index: 12,08644

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR