Diisobutylaluminium hydride 1 M in toluene

Synonyms: Di-i-butylaluminum hydride, DIBAL-H

215007-800ML 215007-100ML 215007-18L
220043-883EA 219.46 USD
220043-883 220043-881 220043-882
Diisobutylaluminium hydride 1 M in toluene
Diisobutylaluminium hydride
Formula: C₈H₁₉Al
MW: 142.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00008928
CAS Number: 1191-15-7
EINECS: 214-729-9
UN: 3399
ADR: 4.3,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR