Diisobutylaluminium hydride 1 M in heptane

Synonyms: Di-i-butylaluminum hydride, DIBAL-H

214965-800ML 214965-100ML
220044-199EA 190.38 USD
220044-199 220044-198
Diisobutylaluminium hydride 1 M in heptane
Diisobutylaluminium hydride
Formula: C₈H₁₉Al
MW: 142.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00008928
CAS Number: 1191-15-7
EINECS: 214-729-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR