Diisobutylaluminium hydride 1 M in cyclohexane

Synonyms: Di-i-butylaluminum hydride, DIBAL-H

214949-800ML 214949-100ML
220044-196EA 252 USD
220044-196 220044-195
Diisobutylaluminium hydride 1 M in cyclohexane
Diisobutylaluminium hydride
Formula: C₈H₁₉Al
MW: 142.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00008928
CAS Number: 1191-15-7
EINECS: 214-729-9
UN: 3399
ADR: 4.3,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR