2-Amino-4-methylbenzonitrile ≥97%

Synonyms: 2-Cyano-5-methylaniline

214280-1G 214280-5G
220042-235EA 67.95 USD
220042-235 220042-236
2-Amino-4-methylbenzonitrile ≥97%
2-Amino-4-methylbenzonitrile
Formula: C₈H₈N₂
MW: 132.16 g/mol
MDL Number: MFCD00173706
CAS Number: 26830-96-6
EINECS: 248-020-0
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR