1,2,3,4,5-Pentamethylcyclopentadiene

Synonyms: Pentamethylcyclopentadiene, 1,2,3,4,5-Pentamethylcyclopenta-1,3-diene, 1,2,3,4,5-Pentamethyl-1,3-cyclopentadiene

214027-25G 214027-5G
220044-065EA 479.23 USD
220044-065 220044-066
1,2,3,4,5-Pentamethylcyclopentadiene
1,2,3,4,5-Pentamethylcyclopentadiene
Formula: C₁₀H₁₆
MW: 136.24 g/mol
Boiling Pt: 58 °C (44 hPa)
Density: 0.87 g/cm³ (20 °C)
Flash Pt: 44 °C
Storage Temperature: Ambient
MDL Number: MFCD00001354
CAS Number: 4045-44-7
EINECS: 223-743-4
UN: 3295
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR