(±)-2-Methyl-1-pentanol ≥99%

Synonyms: 2-Methylamyl alcohol, 2-Methyl-1-pentanol, 2-Propylpropanol

214019-50ML 214019-250ML
220044-064EA 93.37 USD
220044-064 220044-063
(±)-2-Methyl-1-pentanol ≥99%
(±)-2-Methyl-1-pentanol
Formula: C₆H₁₄O
MW: 102.18 g/mol
Boiling Pt: 148 °C (1013 hPa)
Melting Pt: 0 °C
Density: 0.82 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004745
CAS Number: 105-30-6
EINECS: 203-285-1
UN: 1987
ADR: 3,III

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR