D-(-)-Diethyl tartrate ≥99%

Synonyms: (2S,3S)-Diethyl 2,3-dihydroxysuccinate, (S,S)-(-)-Diethyl tartrate, (2S,3S)-(-)-Diethyl tartrate, (-)-Diethyl D-tartrate, (S,S)-(-)-Tartaric acid diethyl ester, D(-)-Tartaric acid diethyl ester

213969-5G 213969-25G 213969-500G
220044-057EA 54.49 USD
220044-057 220044-055 220044-056
D-(-)-Diethyl tartrate ≥99%
D-(-)-Diethyl tartrate
Formula: C₈H₁₄O₆
MW: 206.2 g/mol
Boiling Pt: 280 °C (1013 hPa)
Melting Pt: 17 °C
Density: 1.2 g/cm³ (20 °C)
MDL Number: MFCD00064451
CAS Number: 13811-71-7
EINECS: 237-458-8

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR