Vinyltrimethylsilane

Synonyms: Trimethyl(vinyl)silane

213950-5G 213950-25G 213950-100G
220044-054EA 67.22 USD
220044-054 220044-053 220044-052
Vinyltrimethylsilane
Vinyltrimethylsilane
Formula: C₅H₁₂Si
MW: 100.24 g/mol
Boiling Pt: 55 °C (1013 hPa)
Density: 0.7038 g/cm³ (20 °C)
MDL Number: MFCD00008606
CAS Number: 754-05-2
EINECS: 212-042-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR