(±)-1,2-Dodecanediol ≥90%

Synonyms: (±)-1,2-Dihydroxy dodecane, 1,2-Dihydroxydodecane, 1,2-Dodecanediol

213721-50G 213721-250G
220044-034EA 88.74 USD
220044-034 220044-033
(±)-1,2-Dodecanediol ≥90%
(±)-1,2-Dodecanediol
Formula: C₁₂H₂₆O₂
MW: 202.34 g/mol
MDL Number: MFCD00004726
CAS Number: 1119-87-5
EINECS: 214-289-8

Order Now

Specification Test Results

Assay 90%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR