Selenium trioxide ≥99.8%, ReagentPlus®

Danger

Synonyms: Selenium dioxide

213365-500G 213365-100G
220044-007EA 510.77 USD
220044-007 220044-006
Selenium trioxide ≥99.8%, ReagentPlus®
Selenium trioxide
Formula: O₃Se
MW: 126.96 g/mol
Melting Pt: 117.85…118.85 °C
Density: 3.6 g/cm³ (20 °C)
CAS Number: 13768-86-0
EINECS: 237-385-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR